FreezeBox CF 1.5mL System, Blue, Package includes 1 each of FZB-S1 FreezeBox, CF Freeze Core -18C to -4C, and CM-02 Tube Module for 48 x 1.5ml cryo tubes, 152 x 170 x 123mm, Uniform, low temperature protection for consistent sample results, typically +/-0.1 C
Catalog #: 76480-460
Supplier: CRYSTAL TECHNOLOGY & INDUSTRIES
Customers Who Bought This Also Bought