LTS® Filtered Pipette Tips, Rainin® LTS® Compatible
Catalog #: 76480-430
Supplier: Azer Scientific Inc.
Specifications
- Volume:200 µl
- Tip Color:Clear
- Filter:+
- Sterile:+
- Tip Style:Low retention, filtered
- For Use With:Rainin® LTS® pipettes
Specifications
About this item
Our sterile, 20, 200 and 1000 µl LTS® pippete filter tips are manufactured from super clear high quality polypropylene. The surfaces of the tips are produced through a special process. This process makes the tip inner surface become super hydrophobic, thus significantly reduces sample loss and provides substantially higher reproducibility when working with critical media.
- LTS® compatible
- Low retention for optimal precision and minimal sample loss
- Barrier filter to prevent aerosol contamination
- Certified DNase-/RNase-/pyrogen-/endotoxin-free
Customers Who Bought This Also Bought