Muffle Furnace, 1300, Compact Benchtop, 120V, Ideal for ignition tests, gravimetric analysis, and heat treating steel parts, Feature single set point digital control, Max Temperature: 1100 C, Includes: 6-ft (1.8-m), three-wire power cord with plug, Capacity: 76 cu in
Catalog #: MP30608-020
Supplier: Thermo Fisher Scientific
Customers Who Bought This Also Bought