Mini Gel Electrophoresis B2 System, Gel dimensions: 12 x 14 cm, LxWxH: 24.5 x 18 x 8 cm (931/32 x 7 x 35/32"), Buffer volume: 800 mL, Leak-resistant, gasketed ultraviolet transmission trays allow tape-free casting in the buffer chamber, Heavy-duty, one-piece combs, safety lid
Catalog #: MP27372-256
Supplier: Thermo Fisher Scientific
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.