Hot Plate Stirrer, Top Plate Material: Aluminum, Voltage: 120 V, Includes: USA Plug Type, Body Material: Aluminum, Electrical: 120V, Controller Type: Feedback Control with PID / Scale, Length: 11.42 in, Size: 140 mm dia, Height: 100 mm, Temperature: 50C to 350C
Catalog #: MP10054-160
Supplier: Thermo Fisher Scientific
Customers Who Bought This Also Bought