Hot Plate, RT2 Digital, 50C to 350C, Max. Sample Load: 25 kg, Plug Type: US-Style, Shape: Round, Voltage: 120 V, Body Material: Cast Aluminum, Top Plate Material: Aluminum, Ceramic, Size5.5 in. Dia, Height: 3.94 in, Length: 11.42 in, Width: 6.34 in.
Catalog #: MP10053-862
Supplier: Thermo Fisher Scientific
Customers Who Bought This Also Bought