ContactUsLinkComponent
Wipe, Disposable, Nonwoven, White, 13.75x12in, Excellent strength and absorbency with low-lint properties, Ideal material for heavy-duty cleaning, wiping, degreasing and surface prep, Material: Polypropylene (PP)
Catalog #: 76478-824
Supplier:  Medline
Wipe, Disposable, Nonwoven, White, 13.75x12in, Excellent strength and absorbency with low-lint properties, Ideal material for heavy-duty cleaning, wiping, degreasing and surface prep, Material: Polypropylene (PP)
Catalog #: 76478-824
Supplier:  Medline
Customers Who Bought This Also Bought