ContactUsLinkComponent
Mop Handle with Frame and Lever Lock, for cleanrooms, adjustable from 36in to 61in, Mop head with a lock to keep in a horizontal position when desired, Low weight of only 1.05 lb. Reduces operator work loads on all surfaces, Dimensions: 16in x 3.75in
Catalog #: 76478-816
Supplier:  Medline
Mop Handle with Frame and Lever Lock, for cleanrooms, adjustable from 36in to 61in, Mop head with a lock to keep in a horizontal position when desired, Low weight of only 1.05 lb. Reduces operator work loads on all surfaces, Dimensions: 16in x 3.75in
Catalog #: 76478-816
Supplier:  Medline
Customers Who Bought This Also Bought