Utility Pad, Made of polyester fibers in a open-textured, nonwoven pattern, Abrasive particles are dispersed throughout and bonded to the nonwoven construction with a durable adhesive, Designed for cleaning of floors, baseboards, and corners, Brown, Measure 4.5in x 10in
Catalog #: 76478-740
Supplier: Medline
Customers Who Bought This Also Bought