Floor Pad, UHS burnishing pad with a special binder formulation that allows the pad to effectively remove black marks and restore gloss, Designed for medium to hard floor finishes, For use with machines up to 3,000 RPM, Pink, 20in
Catalog #: 76478-726
Supplier: Medline
Customers Who Bought This Also Bought