ContactUsLinkComponent
Floor Pad, Eco-Prep, designed to be used wet or dry for the removal of floor finish without the use of harsh stripping chemicals, Excellent for repairing acrylic or urethane finishes for recoating, Machine speed: up to 350 RPM, Maroon, 14in x 28in
Catalog #: 76478-706
Supplier:  Medline
Floor Pad, Eco-Prep, designed to be used wet or dry for the removal of floor finish without the use of harsh stripping chemicals, Excellent for repairing acrylic or urethane finishes for recoating, Machine speed: up to 350 RPM, Maroon, 14in x 28in
Catalog #: 76478-706
Supplier:  Medline
Customers Who Bought This Also Bought