ContactUsLinkComponent
Spike S1 Monoclonal antibody, Clone: [HL13402], Host: Rabbit, Reactivity: SARS Coronavirus 2/SARS-CoV-2 (COVID-19), Isotype: IgG, Immunogen: Carrier-protein conjugated synthetic peptide encompassing a sequence within center region of SARS-CoV-2 Spike, Synonyms: 2019-nCoV Spike, Size: 100ul
Catalog #: 76476-406
Supplier:  Genetex
Spike S1 Monoclonal antibody, Clone: [HL13402], Host: Rabbit, Reactivity: SARS Coronavirus 2/SARS-CoV-2 (COVID-19), Isotype: IgG, Immunogen: Carrier-protein conjugated synthetic peptide encompassing a sequence within center region of SARS-CoV-2 Spike, Synonyms: 2019-nCoV Spike, Size: 100ul
Catalog #: 76476-406
Supplier:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.