ContactUsLinkComponent
Spike S1 Monoclonal antibody, Clone: [HL1], Host: Rabbit, Reactivity: SARS Coronavirus 2/SARS-CoV-2 (COVID-19), Isotype: IgG, Immunogen: Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminal region of spike, Synonyms: 2019-nCoV Spike, Size: 100ul
Catalog #: 76476-372
Supplier:  Genetex
Spike S1 Monoclonal antibody, Clone: [HL1], Host: Rabbit, Reactivity: SARS Coronavirus 2/SARS-CoV-2 (COVID-19), Isotype: IgG, Immunogen: Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminal region of spike, Synonyms: 2019-nCoV Spike, Size: 100ul
Catalog #: 76476-372
Supplier:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.