Spike ELISA Pair [1A9 / HL13402], Reactivity: SARS Coronavirus 2/SARS-CoV-2 (COVID-19), Storage: Store as concentrated solution. Centrifuge briefly prior to opening vial, Synonyms: 2019-nCoV Spike, SARS-CoV-2 Spike, COVID-19 Spike
Catalog #: 76476-354
Supplier: Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.