ContactUsLinkComponent
Spike S2 (ECD) protein, mouse IgG Fc tag, Species: SARS Coronavirus 2/SARS-CoV-2 (COVID-19), Purity: >90%, Expression System: HEK293 cells, Form: Liquid, Buffer: PBS, Synonyms: 2019-nCoV Spike, SARS-CoV-2 Spike, COVID-19 Spike, Size: 100uG
Catalog #: 76476-324
Supplier:  Genetex
Spike S2 (ECD) protein, mouse IgG Fc tag, Species: SARS Coronavirus 2/SARS-CoV-2 (COVID-19), Purity: >90%, Expression System: HEK293 cells, Form: Liquid, Buffer: PBS, Synonyms: 2019-nCoV Spike, SARS-CoV-2 Spike, COVID-19 Spike, Size: 100uG
Catalog #: 76476-324
Supplier:  Genetex
Customers Who Bought This Also Bought