ContactUsLinkComponent
ACE2 (ECD) protein, mouse IgG Fc tag (active), Species: Human, Purity: >90%, Expression System: HEK293 cells, Form: Liquid, Buffer: PBS, Synonyms: angiotensin I converting enzyme 2, ACEH, Application: ELISA, Functional Assay, Size: 100uG
Catalog #: 76476-322
Supplier:  Genetex
ACE2 (ECD) protein, mouse IgG Fc tag (active), Species: Human, Purity: >90%, Expression System: HEK293 cells, Form: Liquid, Buffer: PBS, Synonyms: angiotensin I converting enzyme 2, ACEH, Application: ELISA, Functional Assay, Size: 100uG
Catalog #: 76476-322
Supplier:  Genetex
Customers Who Bought This Also Bought