ContactUsLinkComponent
RdRp (nsp12) Polyclonal antibody, Host: Rabbit, Reactivity: SARS Coronavirus 2/SARS-CoV-2 (COVID-19), Isotype: IgG, Immunogen: Carrier-protein conjugated synthetic peptide encompassing a sequence within center region of SARS-CoV-2 RdRp (nsp12), Synonyms: 2019-nCoV RdRp, Application: WB, Size: 100ul
Catalog #: 76476-302
Supplier:  Genetex
RdRp (nsp12) Polyclonal antibody, Host: Rabbit, Reactivity: SARS Coronavirus 2/SARS-CoV-2 (COVID-19), Isotype: IgG, Immunogen: Carrier-protein conjugated synthetic peptide encompassing a sequence within center region of SARS-CoV-2 RdRp (nsp12), Synonyms: 2019-nCoV RdRp, Application: WB, Size: 100ul
Catalog #: 76476-302
Supplier:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.