ContactUsLinkComponent
Spike (D614G Mutant) protein (ECD), His tag (active), Species: SARS Coronavirus 2/SARS-CoV-2 (COVID-19), Purity: > 90% by SDS-PAGE, Expression System: HEK293 cellsRegion/Sequence: C-terminal 8x His tagged; 16-1213aa, MW: 190 kDa, Buffer: PBS, 20% Glycerol, Synonyms: spike glycoprotein, Size: 100uG
Catalog #: 76476-280
Supplier:  Genetex
Spike (D614G Mutant) protein (ECD), His tag (active), Species: SARS Coronavirus 2/SARS-CoV-2 (COVID-19), Purity: > 90% by SDS-PAGE, Expression System: HEK293 cellsRegion/Sequence: C-terminal 8x His tagged; 16-1213aa, MW: 190 kDa, Buffer: PBS, 20% Glycerol, Synonyms: spike glycoprotein, Size: 100uG
Catalog #: 76476-280
Supplier:  Genetex
Customers Who Bought This Also Bought