ContactUsLinkComponent
Kit, Foundation Diamond Plate, 4X36X54in, All components resist corrosion, 2.75in (7 cm) thick tile boasts a load rating of 400 lbs. And is manufactured of 100% recycled polypropylene to stand up to a wide range of chemicals, Structural aluminum frameChemical and corrosion resistant
Catalog #: 76475-138
Supplier:  Wearwell
Kit, Foundation Diamond Plate, 4X36X54in, All components resist corrosion, 2.75in (7 cm) thick tile boasts a load rating of 400 lbs. And is manufactured of 100% recycled polypropylene to stand up to a wide range of chemicals, Structural aluminum frameChemical and corrosion resistant
Catalog #: 76475-138
Supplier:  Wearwell
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.