ContactUsLinkComponent
Tile, Foundation open, 18X18in, to elevate employees to the proper height for optimum health, safety, and performance, 2.75in thick tile boasts a load rating of 400lbs and is manufactured of 100% recycled polypropylene to stand up to a wide range of chemicals, provides undeniable traction
Catalog #: 76474-994
Supplier:  Wearwell
Tile, Foundation open, 18X18in, to elevate employees to the proper height for optimum health, safety, and performance, 2.75in thick tile boasts a load rating of 400lbs and is manufactured of 100% recycled polypropylene to stand up to a wide range of chemicals, provides undeniable traction
Catalog #: 76474-994
Supplier:  Wearwell
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.