Mini LED Light Box, Length: 15.8cm (61/4in ), Width: 15.0cm (5?in ), Height: 1.4cm (1/2in ), Backlight Samples for Easy Viewing, Enhances viewing of transparent and semi-transparent objects such as 96-well plates, microscope slides, Petri dishes, electrophoresis gels, and transparencies
Catalog #: 76475-882
Supplier: Bel-Art Products, a Part of SP
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.