ContactUsLinkComponent
CfDNA/cfRNA Preservation Blood Tube, protected for up to 7 days at 15-25C, preserves integrity of cell-free nucleic acids by prevention of blood cell lysis and releasing cellular nucleic acids, as well as prevention of cell-free nucleic acids degradation, Blood draw: 8.1 mL, Size: 50preps
Catalog #: 76474-280
Supplier:  Zymo Research
CfDNA/cfRNA Preservation Blood Tube, protected for up to 7 days at 15-25C, preserves integrity of cell-free nucleic acids by prevention of blood cell lysis and releasing cellular nucleic acids, as well as prevention of cell-free nucleic acids degradation, Blood draw: 8.1 mL, Size: 50preps
Catalog #: 76474-280
Supplier:  Zymo Research
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.