ContactUsLinkComponent
Spike RBD protein, His tag (active), Species: SARS Coronavirus 2/SARS-CoV-2 (COVID-19), Purity: > 95% by SDS-PAGE, Expression System: HEK293 cells, Lyophilized powder, Buffer: Lyophilized from 0.22 um-filtered PBS (pH 7.4), Synonyms: 2019-nCoV Spike, SARS-CoV-2 Spike, COVID-19 Spike, Size: 100uG
Catalog #: 76476-260
Supplier:  Genetex
Spike RBD protein, His tag (active), Species: SARS Coronavirus 2/SARS-CoV-2 (COVID-19), Purity: > 95% by SDS-PAGE, Expression System: HEK293 cells, Lyophilized powder, Buffer: Lyophilized from 0.22 um-filtered PBS (pH 7.4), Synonyms: 2019-nCoV Spike, SARS-CoV-2 Spike, COVID-19 Spike, Size: 100uG
Catalog #: 76476-260
Supplier:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.