Order Entry
ContactUsLinkComponent
STANDARD PROFESSIONAL PLANT PRESS
Plant Press
Catalog #: 470332-436
Supplier:  Ward's Science
STANDARD PROFESSIONAL PLANT PRESS
Plant Press
Catalog #: 470332-436
Supplier:  Ward's Science

Specifications

  • Description:
    Professional plant press
  • Cat. no.:
    470332-436
  • Length:
    18"
  • Width:
    12"

Specifications

About this item

Larger size for herbarium specimens.

  • Large size for whole specimens
  • Strong straps for even compression

This professional plant press has a solid oak frame that is pinned and glued for maximum strength and durability for years of use. All outside edges are smoothed and beveled to reduce strap wear. Two extra-long straps with buckles (~50" in length) allow for maximum press capacity. The Student press is constructed from two ¼" plywood boards with hardwood strips reinforcing the sides. These hardwood strips are carefully beveled to prevent breakage and provide uniform compression on the specimens.

Delivery: Both presses include straps, 12 ventilators and 12 driers.
Customers Who Bought This Also Bought