Order Entry
ContactUsLinkComponent
Test Strip, Fentanyl Forensic, Single Drug, Accuracy: >98%, Time to result: 5minutes, lateral flow chromatographic immunoassay for qualitative detection of Fentanyl in liquid/powder substances at concentration of 200ng/mL, Content: Test strips, Swabs, Test Tubes, Extraction Buffers, 100/kit
Rapid Response™ Fentanyl Test Strip, BTNX
Catalog #: 76473-480
Supplier:  BTNX INC.
Test Strip, Fentanyl Forensic, Single Drug, Accuracy: >98%, Time to result: 5minutes, lateral flow chromatographic immunoassay for qualitative detection of Fentanyl in liquid/powder substances at concentration of 200ng/mL, Content: Test strips, Swabs, Test Tubes, Extraction Buffers, 100/kit
Rapid Response™ Fentanyl Test Strip, BTNX
Catalog #: 76473-480
Supplier:  BTNX INC.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Fentanyl (FYL) Test Strip
  • Size:
    100 tests

Specifications

About this item

The Rapid Response™ Fentanyl (FYL) Test Strip is a rapid visual immunoassay for the qualitative detection of fentanyl at a cut-off of 200 ng/mL for liquid/powder or 20 ng/mL for urine.

  • Parameter: FYL
  • Calibrator: Fentanyl
  • Cut-off: 200 ng/ml (Liquid/Powder), 20 ng/ml (Urine)

Interpretation of test results are: positive (one line), negative (two lines), invalid (no lines or no control line).

Delivery: Includes test strips, product insert.
Caution: This test does not evaluate drug safety or purity. This test is for forensic use only.
Customers Who Bought This Also Bought