Swab, Sterile, Knit Polyester, Micro, for cleaning broad surfaces and flat areas, Head Width: 3.2mm, Head Thickness: 2.5mm, Head Length: 11mm, Handle Width: 2.2mm, Handle Thickness: 2.2mm, Handle Length: 60mm, Total Swab Length: 70mm
Catalog #: 76472-826
Supplier: Texwipe
Customers Who Bought This Also Bought