Swab, Sterile, Knit Polyester, Long handle, for cleaning broad surfaces and flat areas, Head Width: 7mm, Head Thickness: 2.8mm, Head Length: 16.8mm, Handle Width: 3.2mm, Handle Thickness: 3.2mm, Handle Length: 145.5mm, Total Swab Length: 162.3mm
Catalog #: 76472-822
Supplier: Texwipe
Customers Who Bought This Also Bought