Swab, Sterile, Knitted, Polyester, for cleaning broad surfaces and flat areas, Head Width: 25.30 mm, Head Thickness: 5.25mm, Head Length: 35.5mm, Handle Width: 10mm, Handle Thickness: 3.8mm, Handle Length: 91.5mm, Total Swab Length: 127mm, Flat large head paddle, long handle
Catalog #: 76472-820
Supplier: Texwipe
Customers Who Bought This Also Bought