E 105/1 Quarter basket insert (for up to 200 test tubes 12x165mm, 6 compartments incl. A13 lid
Catalog #: CA76472-980
Supplier: Miele
E 105/1 Quarter basket insert (for up to 200 test tubes 12x165mm, 6 compartments incl. A13 lid
Catalog #: CA76472-980
Supplier: Miele
Customers Who Bought This Also Bought
VWR® THree-Channel Electronic Timer and Clock with Certificate of Calibration
Supplier: VWR International
$142.45 to $2,173.12