ContactUsLinkComponent
PG 8595, Aqua purificator, For two full or partial demineralisation cartridges, Suitable for VE/TE P 2800 & VE/TE P 2000, Possible to hold conductivity meter/flow meter, External dimensions: H 835 (820 w/o lid), W 300, D 600 mm
Catalog #: CA76472-958
Supplier:  Miele
PG 8595, Aqua purificator, For two full or partial demineralisation cartridges, Suitable for VE/TE P 2800 & VE/TE P 2000, Possible to hold conductivity meter/flow meter, External dimensions: H 835 (820 w/o lid), W 300, D 600 mm
Catalog #: CA76472-958
Supplier:  Miele
Customers Who Bought This Also Bought