ContactUsLinkComponent
Module, A 302/2, for optimum loading of laboratory glassware, can be equipped with 32 nozzles, Suitable for lab, Glassware such as Erlenmeyer and round flasks, Module without injector nozzles, Part of the EasyLoad system, Nozzle spacing 60 mm across the width, 57 mm across the depth
Catalog #: CA76415-394
Supplier:  Miele
Module, A 302/2, for optimum loading of laboratory glassware, can be equipped with 32 nozzles, Suitable for lab, Glassware such as Erlenmeyer and round flasks, Module without injector nozzles, Part of the EasyLoad system, Nozzle spacing 60 mm across the width, 57 mm across the depth
Catalog #: CA76415-394
Supplier:  Miele
Customers Who Bought This Also Bought