ContactUsLinkComponent
Module, A 301/4, for optimum loading of laboratory glassware, can be equipped with 18 nozzles, Suitable for lab, Glassware such as Erlenmeyer and round flask, Module without injector nozzles, Part of the EasyLoad system, Nozzle spacing 157 mm across the width, 78 mm across the depth
Catalog #: CA76415-392
Supplier:  Miele
Module, A 301/4, for optimum loading of laboratory glassware, can be equipped with 18 nozzles, Suitable for lab, Glassware such as Erlenmeyer and round flask, Module without injector nozzles, Part of the EasyLoad system, Nozzle spacing 157 mm across the width, 78 mm across the depth
Catalog #: CA76415-392
Supplier:  Miele
Customers Who Bought This Also Bought