ContactUsLinkComponent
Module, A 302/5, for optimum loading of laboratory glassware, can be equipped with 18 nozzles, Suitable for lab, Glassware such as Erlenmeyer and round flasks, Equipped with 9 A 842 and 9 A 843 nozzles, Part of the EasyLoad system, Nozzle spacing 78 mm across the width, 157 mm across the depth
Catalog #: CA76318-276
Supplier:  Miele
Module, A 302/5, for optimum loading of laboratory glassware, can be equipped with 18 nozzles, Suitable for lab, Glassware such as Erlenmeyer and round flasks, Equipped with 9 A 842 and 9 A 843 nozzles, Part of the EasyLoad system, Nozzle spacing 78 mm across the width, 157 mm across the depth
Catalog #: CA76318-276
Supplier:  Miele
Customers Who Bought This Also Bought