ContactUsLinkComponent
Module, A 302/3, for optimum loading of laboratory glassware, can be equipped with 8 nozzles, Suitable for lab, Glassware such as Erlenmeyer and round flasks, Equipped with 4 A 840 and 4 A 841 nozzles, Part of the EasyLoad system, Nozzle spacing 118 mm across the width, 118 mm across the depth
Catalog #: CA76318-274
Supplier:  Miele
Module, A 302/3, for optimum loading of laboratory glassware, can be equipped with 8 nozzles, Suitable for lab, Glassware such as Erlenmeyer and round flasks, Equipped with 4 A 840 and 4 A 841 nozzles, Part of the EasyLoad system, Nozzle spacing 118 mm across the width, 118 mm across the depth
Catalog #: CA76318-274
Supplier:  Miele
Customers Who Bought This Also Bought