Chiller, recirculating, non-standard voltage; Up to 900 watts (60Hz) 750 watts (50Hz) Air Cooled, Basic Controller, T1 Pump option are now available in low noise models. Quiet performance (<58 dBA) promotes a safe and reduced stress environment in your lab. 5C to 40C Temperature Range
Catalog #: 76472-738
Supplier: Thermo Fisher Scientific
Customers Who Bought This Also Bought