Cartridge, Digital Dispenser, 8-ch for Multidrop Pico 8, Voltage: 100 to 240V, Dispensing Range11 pL to 20 uL (8towell Cartridge), 1 nL to 200 uL (4towell Cartridge), For Use With Drug combination studies, Biochemical assays, Cell-based assays, Dimensions (L x W x H): 8x10.5x2.5 cm, Volume: 20 uL
Catalog #: 76473-018
Supplier: Thermo Fisher Scientific
Customers Who Bought This Also Bought