Cartridge, Digital Dispenser, 1-ch for Multidrop Pico 1, Voltage: 100 to 240V, Dispensing Range11 pL to 20 uL or 1 nL to 200 uL, For Use With Drug combination studies, Biochemical assays, Cell-based assays, Dimensions (L x W x H):8.2 x 6.5 X 2.2 cm, Volume: 20 uL
Catalog #: 76473-012
Supplier: Thermo Fisher Scientific
Customers Who Bought This Also Bought