Digital Dispenser, Pico 8, Voltage: 100 to 240V 50/60Hz, For Use With Drug combination studies, Biochemical assays, Cell-based assays, Interface: USB, Dispensing Range: 11pL to 20uL (8 well Cartridge), 1nL to 200uL (4 well Cartridge), Dimensions (L x W x H): 47 cm x 38 cm x 23 cm
Catalog #: 76473-010
Supplier: Thermo Fisher Scientific
Customers Who Bought This Also Bought