ContactUsLinkComponent
Kit, GFX PCR DNA and Gel Band Purification, Designed for the rapid purification and concentration of PCR products or DNA fragments ranging in size from 50 bp to 10 kb, used to purify DNA from reaction volumes up to 100 ul or agarose gel slices up to 900 mg, Size: 100rxns
Catalog #: MPCA29648122
Supplier:  Cytiva
Kit, GFX PCR DNA and Gel Band Purification, Designed for the rapid purification and concentration of PCR products or DNA fragments ranging in size from 50 bp to 10 kb, used to purify DNA from reaction volumes up to 100 ul or agarose gel slices up to 900 mg, Size: 100rxns
Catalog #: MPCA29648122
Supplier:  Cytiva
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.