ContactUsLinkComponent
Column, NAP-5, disposable, For rapid and efficient purification of DNA and oligonucleotides (10-mers) in less than 15 min by gravity flowPrepacked with Sephadex G-25 DNA Grade in distilled water containing 0.15% Kathon CG/ICP Biocide, Includes 50 columns, Size: 0.5ml
Catalog #: MPCA29648118
Supplier:  Cytiva
Column, NAP-5, disposable, For rapid and efficient purification of DNA and oligonucleotides (10-mers) in less than 15 min by gravity flowPrepacked with Sephadex G-25 DNA Grade in distilled water containing 0.15% Kathon CG/ICP Biocide, Includes 50 columns, Size: 0.5ml
Catalog #: MPCA29648118
Supplier:  Cytiva
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.