ContactUsLinkComponent
PuReTaq Ready-To-Go PCR Bead, (0.2 ml tubes), Includes: 2.5 units of recombinant PuReTaq DNA polymerase, dATP, dCTP, dGTP, dTTP, stabilizers, BSA and reaction buffer, concentration of each dNTP is 200 uM in 10 mM Tris-HCl (pH 9.0 at room temperature), 50 mM KCl and 1.5 mM MgCl2, Size: 96reactions
Catalog #: MPCA29648117
Supplier:  Cytiva
PuReTaq Ready-To-Go PCR Bead, (0.2 ml tubes), Includes: 2.5 units of recombinant PuReTaq DNA polymerase, dATP, dCTP, dGTP, dTTP, stabilizers, BSA and reaction buffer, concentration of each dNTP is 200 uM in 10 mM Tris-HCl (pH 9.0 at room temperature), 50 mM KCl and 1.5 mM MgCl2, Size: 96reactions
Catalog #: MPCA29648117
Supplier:  Cytiva
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.