ContactUsLinkComponent
Kit, TempliPhi 100 Amplification, Used to amplify single- or double-stranded circular DNA using Phi29 DNA polymerase rolling circle amplification (RCA), Size: 100reactions
Catalog #: MPCA29648116
Supplier:  Cytiva
Kit, TempliPhi 100 Amplification, Used to amplify single- or double-stranded circular DNA using Phi29 DNA polymerase rolling circle amplification (RCA), Size: 100reactions
Catalog #: MPCA29648116
Supplier:  Cytiva
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.