ContactUsLinkComponent
Marking Tape, Black/Yellow Diagonal Stripes, 2inx54 Ft, Adhesive Vinyl, Flexible vinyl with acrylic adhesive, Non-glare surface printed, Moisture resistant, Service temperature range: -40 to 176F
Floor Marking Tape, Black/Yellow, Accuform
Catalog #: 76471-366
Marking Tape, Black/Yellow Diagonal Stripes, 2inx54 Ft, Adhesive Vinyl, Flexible vinyl with acrylic adhesive, Non-glare surface printed, Moisture resistant, Service temperature range: -40 to 176F
Floor Marking Tape, Black/Yellow, Accuform
Catalog #: 76471-366
Specifications
  • Length:
    54'
  • Width:
    2"
  • Color:
    Black/Yellow
  • Description:
    Floor marking tape
  • Cat. no.:
    76471-366
Specifications
About this item
Marking tape is used to mark specific areas and promote safety by identifying potential hazard areas.
  • Vinyl adheres to floors, walls, piping, posts, beams, handrails, stairways, machinery, and equipment
  • Permanent self-adhesive back with no liner
Extremely durable floor marking tape is waterproof and wear-resistant, while offering minor resistance to acids, oils, grease, caustics, alkalis, and most solvents.
Customers Who Bought This Also Bought