ContactUsLinkComponent
Pipette, adjustable volume, 12-channel, M12-200, Autoclavable at 121C (2bar), Non-sterile, Material: Polypropylene, Length (Metric): 23.7cm, Height (Metric): 12.3cm, Width (Metric): 14cm, Low operating forces and a short stroke ensure relaxed pipetting, Universal tip cone, Volume (Metric): 20-200 UL
Catalog #: MP76383-114
Supplier:  Brandtech
Pipette, adjustable volume, 12-channel, M12-200, Autoclavable at 121C (2bar), Non-sterile, Material: Polypropylene, Length (Metric): 23.7cm, Height (Metric): 12.3cm, Width (Metric): 14cm, Low operating forces and a short stroke ensure relaxed pipetting, Universal tip cone, Volume (Metric): 20-200 UL
Catalog #: MP76383-114
Supplier:  Brandtech
Customers Who Bought This Also Bought