ContactUsLinkComponent
Pipette, adjustable volume, 8-channel, M8-300, Autoclavable at 121C (2bar), Non-sterile, Material: Polypropylene, Length (Metric): 23.7cm, Height (Metric): 12.3 cm, Width (Metric): 14 cm, Low operating forces and a short stroke ensure relaxed pipetting, Universal tip cone, Volume (Metric): 30-300 UL
Catalog #: MP76383-102
Supplier:  Brandtech
Pipette, adjustable volume, 8-channel, M8-300, Autoclavable at 121C (2bar), Non-sterile, Material: Polypropylene, Length (Metric): 23.7cm, Height (Metric): 12.3 cm, Width (Metric): 14 cm, Low operating forces and a short stroke ensure relaxed pipetting, Universal tip cone, Volume (Metric): 30-300 UL
Catalog #: MP76383-102
Supplier:  Brandtech
Customers Who Bought This Also Bought