Pipette, electronic, Display: LCD, Battery Type: Rechargeable, Number of Channels: 12, Finger Style: Ergonomic, Includes: Battery, power supply unit, 120VAC adapter with charging cable, one pipette tip sample, pipette stand, tip-box SL filled with appropriate tip, Volume (Metric): 0.5 to 10 uL
Catalog #: MP18900-522
Supplier: Brandtech