ContactUsLinkComponent
PH Indicator Strip, PH Range: 0 - 13, Instant pH readings, Accurate for a wide range of routine pH testing, Convenient and portable for field use, have been impregnated with a mixture of several indicators. On contact with the sample solution they assume a particular color, 6x85mm
Catalog #: MPCA29648153
Supplier:  Cytiva
PH Indicator Strip, PH Range: 0 - 13, Instant pH readings, Accurate for a wide range of routine pH testing, Convenient and portable for field use, have been impregnated with a mixture of several indicators. On contact with the sample solution they assume a particular color, 6x85mm
Catalog #: MPCA29648153
Supplier:  Cytiva