ContactUsLinkComponent
Syringeless Filter, Mini-UniPrep, std cap, Color: Translucent, Pore Size: 0.2 um, Filter media: Polyvinylidene Difluoride (PVDF), integrate an autosampler vial, filtration membrane, plunger, and cap/septa, For HPLC/UHPLC sample preparation, Compatible with most major autosamplers
Catalog #: MPCA29648147
Supplier:  Cytiva
Syringeless Filter, Mini-UniPrep, std cap, Color: Translucent, Pore Size: 0.2 um, Filter media: Polyvinylidene Difluoride (PVDF), integrate an autosampler vial, filtration membrane, plunger, and cap/septa, For HPLC/UHPLC sample preparation, Compatible with most major autosamplers
Catalog #: MPCA29648147
Supplier:  Cytiva