Water bath, Shallow, Model: precision gp 2S, Chamber Material: Stainless Steel, Voltage: 120/230V, Heating Capacity: 300 w, Material: Epoxy Powder-coated Steel, Includes: Clear Polycarbonate Gable Cover and Rubber Duck, Dimensions (L x W x H): 9.7 x 14 x 9.1 in. (246 x 355 x 232 mm), Size: 2l
Catalog #: MPTSGP2SPM
Supplier: Thermo Fisher Scientific
Customers Who Bought This Also Bought