Water Purification System, Type 1, xCAD Plus UV/UF (wall version), Voltage: 90/240 V, Feedwater Connector: 3/4 in, Integrated, with remote dispenser, Temperature Range (Metric): 2C to 35C, Total Organic Carbon: 1 to 5 ppb, Operating Pressure: 0.1 to 6 bar, Dimension: 615 mmx372 mmx330 mm
Catalog #: MPCA89414-732
Supplier: Thermo Fisher Scientific
Customers Who Bought This Also Bought